Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD108 Mouse, anti-Human, R718, Clone: KS-2, BD OptiBuild™

Catalog No. BDB752274 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752274 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752274 Supplier BD Biosciences Supplier No. 752274
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The KS-2 monoclonal antibody specifically recognizes the John-Milton-Hagen (JMH) blood group antigen. CD108 is a 76-80 kDa glycosylphosphatidylinositol (GPI)-linked protein. It is expressed on erythrocytes, at low levels on lymphocytes, but higher levels are detected on lymphoblasts and lymphoblastic cell lines. An adhesion function for CD108 has been suggested but this has not been demonstrated. Autoantibodies to this molecule are not uncommon and often associated with the loss of JMH antigen expression on erythrocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD108
Applications Flow Cytometry
Classification Monoclonal
Clone KS-2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.