Learn More
Invitrogen™ CD109 Monoclonal Antibody (HU17), PE, eBioscience™

Description
Description: The HU17 monoclonal antibody reacts with human CD109, also known as the platelet-specific Gov antigen. CD109, a GPI-anchored monomeric glycoprotein, exists in two isoforms of 120 and 150 kDa, which are proteolytically degraded from a 170 kDa form. CD109 is expressed on activated T cells, activated platelets and endothelial cells. CD109 expression is also found on a subset of CD34+ cells in adult and fetal bone marrow which contain megakaryocyte, myeloid, erythroid and multilineage progenitors, as well as pluriopotent hematopoietic stem cells. Recently, it has been demonstrated that CD109 is expressed on a variety of human cancer cell lines, and also lung and esophogeal cell carcinomas. In addition, CD109 was recently found to be a negative regulator of TGF-beta in keratinocytes. Applications Reported: This HU17 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HU17 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD109 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HU17 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CD109 |
| Gene Accession No. | Q6YHK3 |
| Gene Alias | 150 kDa TGF-beta-1-binding protein; 9930012E15Rik; activated T-cell marker CD109; AI480638; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; Cd109; CD109 antigen; CD109 molecule; CPAMD7; GARP; Gov platelet alloantigens; GPI-anchored alpha 2 macroglobulin-related protein; GPI-anchored alpha-2 macroglobulin-related protein; p180; platelet-specific Gov antigen; r150; RP11-525G3.1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.