Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD112 (Nectin-2) Monoclonal Antibody (R2.477), APC, eBioscience™
GREENER_CHOICE

Catalog No. 501123070
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-3070 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-3070 Supplier Invitrogen™ Supplier No. 17112842
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This R2.477 monoclonal antibody reacts with human CD112 (also known as Nectin-2 or Poliovirus Receptor Related 2), a 60-65 kDa immunoglobulin-like transmembrane cell adhesion molecule. This cell surface molecule is expressed on hematopoietic cells, endothelial, epithelial, as well as tumor cell lines such as Jurkat, U937, and MCF7. CD112 is a ligand for DNAM-1 (CD226), a leukocyte adhesion molecule expressed on natural killer (NK) cells. Interaction between CD112 and DNAM-1 promotes NK cytotoxic activity, leading to tumor cell lysis. In addition to its role in tumor suppression, CD112 plays a cooperative role in cell-cell adhesion, particularly in the formation of adherens junctions between neighboring epithelial cells and fibroblasts. CD112 can form homophilic and heterophilic interactions to regulate contact inhibition of cell movement and proliferation. Finally, CD112 has been reported to be a receptor for herpesvirus entry into cells. This monoclonal antibody has been reported to block cell adhesion and herpesvirus entry. Applications Reported: This R2.477 antibody has been reported for use in flow cytometric analysis. Applications Tested: This R2.477 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirical...

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD112 (Nectin-2)
Applications Flow Cytometry
Classification Monoclonal
Clone R2.477
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene NECTIN2
Gene Accession No. Q92692
Gene Alias AI325026; AI987993; CD112; herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; MPH; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs
Gene Symbols NECTIN2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5819
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.