Learn More
ACROBiosystems Recombinant Protein;Human CD117 / c-kit Protein, Fc Tag;HEK293;1MG;CD7,CD117,SCFR,c-Kit,KIT
Supplier:  ACROBiosystems CD7H52551MG
Encompass_Preferred
Recombinant Protein;Human CD117 / c-kit Protein, Fc Tag;1MG;Human CD117, Fc Tag (CD7-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession # P10721-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81.9 kDa. The protein migrates as 100-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD117, Fc Tag (Cat. No. CD7-H5255) at 2 μg/mL (100 μl/well) can bind Mouse SCF, His Tag (Cat. No. SCF-M5228) with a linear range of 0.05-1.2 ng/mL (Routinely tested).;Immobilized Human CD117, Fc Tag (Cat. No. CD7-H5255) at 2 μg/mL (100 μl/well) can bind Biotinylated Mouse SCF (Cat. No. SCF-M8228) with a linear range of 0.25-20 ng/mL (QC tested).
Catalog No. 50-165-8968
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.