Learn More
Invitrogen™ CD11a (LFA-1alpha) Monoclonal Antibody (BL1H8), FITC

Description
Mouse anti Pig CD11a, clone BL1H8 recognizes porcine CD11a, binding to a conformational epitope on the alpha chain of LFA-1 (CD11a/CD18), expressed on a range of cells including peripheral blood lymphocytes, monocytes, granulocytes and alveolar macrophages.
Specifications
Specifications
| Antigen | CD11a (LFA-1alpha) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | BL1H8 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 1% BSA and 0.09% sodium azide |
| Gene | ITGAL |
| Gene Accession No. | 0 |
| Gene Alias | (p180); alpha L integrin; antigen CD11A (p180); antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; antigen CD11A p180; CD11 antigen-like family member A; CD11a; integrin alpha L; integrin alphaL; integrin alpha-L; integrin alpha-L precursor; integrin gene promoter; integrin subunit alpha L; integrin surface receptor; integrin, alpha L; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); ITGAL; Leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte function-associated molecule 1 alpha chain; LFA-1; LFA-1 alpha; LFA1A; LFA-1A; Ly-15; Ly-21; lymphocyte antigen 15; lymphocyte function associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen 1; lymphocyte function-associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.