Learn More
Invitrogen™ CD11a Monoclonal Antibody (MEM-25), FITC

Description
This antibody reacts with an extracellular epitope of CD11a (alpha subunit of human LFA-1), a 170-180 kDa type I transmembrane glycoprotein expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils and eosinophils.
Specifications
Specifications
| Antigen | CD11a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM-25 |
| Concentration | 0.01 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | ITGAL |
| Gene Accession No. | P20701 |
| Gene Alias | (p180); alpha L integrin; antigen CD11A (p180); antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; antigen CD11A p180; CD11 antigen-like family member A; CD11a; integrin alpha L; integrin alphaL; integrin alpha-L; integrin alpha-L precursor; integrin gene promoter; integrin subunit alpha L; integrin surface receptor; integrin, alpha L; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); ITGAL; Leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte function-associated molecule 1 alpha chain; LFA-1; LFA-1 alpha; LFA1A; LFA-1A; Ly-15; Ly-21; lymphocyte antigen 15; lymphocyte function associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen 1; lymphocyte function-associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.