Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1089474A70
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1089474A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1089474A70 Supplier R&D Systems Supplier No. NB11089474AF750

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

CD11b Polyclonal specifically detects CD11b in Bovine, C. elegans, Chinese Hamster, Human, Monkey, Mouse, Rat, Rhesus Macaque samples. It is validated for Dual RNAScope ISH-IHC, Flow Cytometry, Flow Cytometry (Cell Surface), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In-situ Hybridization, Single Cell Western, Western Blot.

Specifications

Antigen CD11b
Applications Dual RNAScope ISH-IHC, Flow Cytometry, Flow Cytometry (Cell Surface), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), Single Cell Western, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement
Gene Symbols ITGAM
Host Species Rabbit
Immunogen Rabbit Polyclonal CD11b Antibody was made to a synthetic peptide within residues 250-350 of the mouse CD11b protein. [Swiss-Prot# P05555]
Molecular Weight of Antigen 127.2 kDa
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3684
Target Species Bovine, C. elegans, Chinese Hamster, Human, Monkey, Mouse, Rat, Rhesus Macaque
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.