Learn More
Invitrogen™ CD11c Monoclonal Antibody (FK24)

Description
The antibody reacts with the alpha chain the 150 kD molecule of the CD11c/CD18 Integrin complex (g150/95) which is expressed on monocytes and in a lower density on granulocytes and NK-cells of peripheral blood. It also reacts with many myelomonocytic and monocytic leukemias and (weakly) with part of the T8 positive cells (OKM1-like). The Integrins are important molecules in cellular adhesion and cellular interactions. Positive controle perifere blood lymphocytes.
Specifications
Specifications
| Antigen | CD11c |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | FK24 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide |
| Gene | Itgax |
| Gene Accession No. | P20702 |
| Gene Alias | AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.