Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD11c Monoclonal Antibody (N418), Brilliant Ultra Violet™ 563, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 365011482
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
365-0114-82 100 μg
365-0114-80 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 365-0114-82 Supplier Invitrogen™ Supplier No. 365011482
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Applications Flow Cytometry
Classification Monoclonal
Clone N418
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 563
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Itgax
Gene Accession No. Q9QXH4
Gene Alias AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6
Gene Symbols Itgax
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16411
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.