Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c Mouse anti-Human, Rhesus, Alexa Fluor 647, Clone: 3.9, BD Biosciences™

Catalog No. p-7231838 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565911 25 μg
BDB565912 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB565911 Supplier BD Biosciences Supplier No. 565911
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD11c Monoclonal antibody specifically detects CD11c in Human, Rhesus samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Applications Flow Cytometry
Classification Monoclonal
Clone 3.9
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ITGAX; AlphaX Integrin; Axb2; Integrin alpha-X; CR4; SLEB6; p150,95 alpha
Host Species Mouse
Immunogen Human monocytes and synovial cells
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.