Learn More
Invitrogen™ CD120a (TNF Receptor I) Monoclonal Antibody (55R-170), Functional Grade, eBioscience™, Invitrogen™
Description
Description: The 55R-170 monoclonal antibody reacts with mouse Tumor Necrosis Factor Receptor I (TNFRI, TNFR-p55, TNFRSF1A, CD120a). TNFRI is expressed in a variety of cell types. The soluble TNF receptors are truncated forms of cell surface receptors with neutralizing activity on both TNF-alpha and TNF-beta. It has been suggested that the proinflammatory and immunosuppressive properties of TNF segregate at the level of its receptors and that the pool of TNF bound to soluble receptors could represent a reservoir to control the TNF release. Applications Reported: The 55R-170 antibody has been reported for use in ELISA, neutralization of mouse TNFR I bioactivity and flow cytometric analysis. Applications Tested: The Functional Grade Purified 55R-170 antibody has been tested by LAL assay to verify low endotoxin levels and in bioassay for neutralization of mouse TNFR I bioactivity. The 55R-170 antibody at 25 μg/mL has been found to inhibit by 50% the biological effects of 1 ng/mL mouse TNF alpha in an assay of TNF alpha induced cytotoxicity of L929 cells. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in the Best Protocols™ section. Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE.
Specifications
Specifications
| Antigen | CD120a (TNF Receptor I) |
| Applications | Functional Assay, Neutralization |
| Classification | Monoclonal |
| Clone | 55R-170 |
| Concentration | 1 mg/mL |
| Conjugate | Functional Grade |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | Tnfrsf1a |
| Gene Accession No. | P25118 |
| Gene Alias | CD120a; CD120a antigen; FPF; hypothetical protein; metalloproteinase inhibitor 4; MGC19588; MS5; n-smase activation assoc. factor; p55; p55 TNF receptor; p55-R; p60; p62; p63; sCD120a; soluble CD120a; soluble TNFR1; sTNF RI; sTNFR I; sTNFR1; sTNFRI; TBP1; TBPI; TIMP metallopeptidase inhibitor 4; Timp4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4; TNF receptor alpha chain; TNF receptor superfamily member 1A; TNF-alphaR1; TNF-alpha-R1; TNFalpha-R1; TNFAR; TNFR; TNF-R; TNFR I; TNFR1; TNF-R1; Tnfr-1; TNFR1-d2; Tnfr-2; TNFR55; TNF-R55; TNFR60; TNFRI; TNF-RI; TNF-R-I; TNFR-I; TNFRp55; Tnfrsf1a; tumor necrosis factor binding protein 1; Tumor necrosis factor receptor; tumor necrosis factor receptor 1; tumor necrosis factor receptor 1A isoform beta; tumor necrosis factor receptor superfamily member 1A; Tumor necrosis factor receptor superfamily member 1A, membrane form; tumor necrosis factor receptor superfamily, member 1A; tumor necrosis factor receptor type 1; tumor necrosis factor receptor type I; tumor necrosis factor-alpha receptor; Tumor necrosis factor-binding protein 1 |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.