Learn More
Invitrogen™ CD120b (TNF Receptor II) Monoclonal Antibody (TR75-54), eBioscience™, Invitrogen™

Description
Description: The TR75-54 monoclonal antibody reacts with mouse Tumor Necrosis Factor Receptor II (TNFRII, TNFR-p80, TNFRSF1B, CD120b). TNFRII is expressed in a variety of cell types and strongly expressed on stimulated T and B lymphocytes. TNFRII is the main TNF receptor found on circulating T cells and is the major mediator of autoregulatory apoptosis in CD8+ cells. The soluble TNF receptors are truncated forms of cell surface receptors with neutralizing activity on both TNF-alpha and TNF-beta. It has been suggested that the proinflammatory and immunosuppressive properties of TNF segregate at the level of its receptors and that the pool of TNF bound to soluble receptors could represent a reservoir to control the TNF release. Applications Reported: This TR75-54 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and ELISA. Applications Tested: The TR75-54 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse TNFRII in combination with the biotin TR75-32 (cat. 13-1204) antibody for detection and recombit mouse TNFRII as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL. A standard curve consisting of doubling dilutions of the recombit standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE.
Specifications
Specifications
| Antigen | CD120b (TNF Receptor II) |
| Applications | ELISA, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | TR75-54 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Tnfrsf1b |
| Gene Accession No. | P25119 |
| Gene Alias | CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1B; Tumor necrosis factor receptor superfamily member 1b, membrane form; tumor necrosis factor receptor superfamily, member 1B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2 |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.