Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD123 Monoclonal Antibody (6H6), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™

Catalog No. 502550855
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-0855 25 Tests
50-255-0854 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-255-0855 Supplier Invitrogen™ Supplier No. 367123941
  • $224.00
    $217.65 / Each of 1

    Save $6.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD123
Applications Flow Cytometry
Classification Monoclonal
Clone 6H6
Concentration 5 μL/Test
Conjugate Brilliant Ultraviolet 737
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IL3RA
Gene Accession No. P26951
Gene Alias CD123; CD123 antigen; CDw123; Cyrl; cytokine receptor-like protein CYRL; hIL-3Ra; Il-3 alpha subunit; IL-3 receptor alpha chain; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL3R; IL-3R subunit alpha; Il3ra; IL-3RA; IL-3R-alpha; IL3RAY; IL3RX; IL3RY; interleukin 3 receptor subunit alpha; interleukin 3 receptor, alpha; interleukin 3 receptor, alpha (low affinity); interleukin 3 receptor, alpha chain; interleukin-3 receptor class II alpha chain; Interleukin-3 receptor subunit alpha; Sut-1
Gene Symbols IL3RA
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3563
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.