Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD124 Monoclonal Antibody (X2/45-12), eBioscience™
GREENER_CHOICE

Catalog No. 5012854
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-128-54 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-128-54 Supplier Invitrogen™ Supplier No. 14124982
  • $280.00
    $210.65 / Each of 1

    Save $69.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The monoclonal antibody X2/45-12 reacts with human CD124, which is also known as the IL-4R alpha chain. CD124 can associate with the common gamma chain to form the type I IL-4R or with IL-13Ralpha1 to form the type II IL-4R. While both receptor types can bind IL-4, only the type II receptor can associate with IL-13. The type I IL-4 receptor is found on T, B, NK, mast cells, basophils, and macrophages. The type II IL-4 receptor is found on many non-hematopoietic cells and macrophages. CD124 contains an ITIM that associates with and activates STAT6. IL-4 is responsible for the generation of high-affinity IgE antibodies and driving the polarization of naive Th0 cells into Th2 cells. In mucosal barriers, the cytokine is involved in the recruitment of innate and adaptive immune effector cells and mucous overproduction. Finally, IL-4R also exists as a soluble form that serves primarily as a carrier protein for IL-4 that increases its half-life. The X2/45 antibody has been reported to have antagonist activity. Applications Reported: This X2/45-12 antibody has been reported for use in flow cytometric analysis, and western blotting. (Please use Functional Grade purified X2/45-12, Product # 16-1249-82, in functional assays). Applications Tested: This X2/45-12 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will...

IL4 Receptor alpha (IL4RA) is the alpha chain of the interleukin 4 receptor which binds to both interleukin 4 and interleukin 13 to regulate IgE production, chemokine and mucus production at sites of allergic inflammmation. The IL4 response is involved in promoting Th2 differentiation. The secreted extracellular domain of IL4R alpha, called sIL4R alpha, can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. It has no signaling abilities. Allelic variations in the IL4RA gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitus, asthma, or eczema.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD124
Applications Flow Cytometry, Functional Assay, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone X2/45-12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IL4R
Gene Accession No. P24394
Gene Alias 582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4R subunit alpha; IL4RA; IL-4RA; IL-4R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha
Gene Symbols IL4R
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3566
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.