Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD126 Monoclonal Antibody (D7715A7), PE-Cyanine7, eBioscience™
GREENER_CHOICE

Catalog No. 25126182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-126-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-126-182 Supplier Invitrogen™ Supplier No. 25126182
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody D7715A7 recognizes mouse CD126, the IL-6 receptor alpha chain, a member of the Ig superfamily. CD126 can heterodimerize with CD130 (gp130) to form the high affinity IL-6 receptor. Once IL-6 is bound, signaling from the CD130 subunit is initiated and the receptor complex is internalized. The IL-6 receptor is also reported to exist as a soluble version, which bound to IL-6 acts in trans to signal. Expression is found on T lymphocytes in the periphery and during development in the thymus is restricted to SP populations, unlike gp130 which is expressed in all thymocytes. Additional expression has been shown on activated B lymphocytes, macrophages, granulocytes and CD34+ hematopoietic progenitor cells. The monoclonal antibody D7715A7 binding can be blocked addition of ligand (IL-6). Applications Reported: This D7715A7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This D7715A7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please prote...

Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD126
Applications Flow Cytometry
Classification Monoclonal
Clone D7715A7
Concentration 0.2 mg/mL
Conjugate PE-Cyanine7
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Il6ra
Gene Accession No. P22272
Gene Alias CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6R; IL-6R 1; IL-6R subunit alpha; IL6R1; IL-6R-1; Il6ra; IL-6RA; IL-6R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991
Gene Symbols Il6ra
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16194
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.