Learn More
Proteintech Group Inc CD133 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 184701AP150UL
Encompass_Preferred
The CD133 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD133. This antibody recognizes human, mouse, rat antigen. The CD133 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Catalog No. 50-225-6419
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.