Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Antibody (M5E2), Alexa Fluor™ 700, R&D Systems™
SDP

Catalog No. NB10077758Z
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10077758Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10077758Z Supplier R&D Systems Supplier No. NB10077758AF700

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

CD14 Monoclonal specifically detects CD14 in Bovine, Canine, Human, Pig, Primate samples. It is validated for Block/Neutralize, CyTOF, Dual RNAScope ISH-IHC, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen CD14
Applications Blocking Assay, CyTOF, Dual RNAScope ISH-IHC, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone M5E2
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Gene Symbols CD14
Host Species Mouse
Immunogen Human peripheral blood cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 929
Test Specificity CD14 (M5E2). Expected to cross react with Chimpanzee, Cynomolgus, Rhesus, Pigtailed Macaque, Capuchin Monkey, Cotton-topped Tamarin, Common Marmoset, Squirrel Monkey.
Target Species Bovine, Canine, Human, Pig, Primate
Content And Storage Store at 4C in the dark.
Product Type Antibody
Isotype IgG2a κ
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.