Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD14 Antibody Pair [HRP]
SDP

Catalog No. NB009888 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009888 5 Plates
NB009887 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB009888 Supplier Novus Biologicals™ Supplier No. NBP2794245PLATES
Add to Cart
Add to Cart

Monoclonal Antibody Pair

specifically detects in Mouse samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 78.1 pg/mL
Assay Sensitivity 78.13-5000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse CD14.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Gene ID (Entrez) 929
Gene Symbol CD14
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 90 ng of recombinant mouse CD14.
  • Capture Antibody: 0.5 mg/mL of mouse anti-mouse CD14 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL rabbit anti-CD14 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target CD14
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 5 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.