Learn More
ABclonal Technology CD141/Thrombomodulin Rabbit
SDP
Supplier:  ABclonal Technology A22989PM100UL
Encompass_Preferred
The protein encoded by this intronless gene is an endothelial-specific type I membrane receptor that binds thrombin This binding results in the activation of protein C which degrades clotting factors Va and VIIIa and reduces the amount of thrombin generated Mutations in this gene are a cause of thromboembolic disease also known as inherited thrombophilia
Catalog No. 50-000-06652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.