Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD148 Monoclonal Antibody (10)
GREENER_CHOICE

Catalog No. PIMA531139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531139 Supplier Invitrogen™ Supplier No. MA531139
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DEP1/PTPRJ.

CD148 is a highly glycosylated transmembrane protein of up to 250 kDa, containing eight fibronectin type III domains in the extracellular region and one tyrosine phosphatase domain in the cytoplasmic region. Expression is found broadly on fibroblasts, epithelial cells, lymphocytes (B, T and NK cells), and myeloid cells (monocytes and granulocytes). CD148 can dephosphorylate proteins such as LAT and PLC-gamma, and is involved in the downregulation of T cell activation. Conversely, CD148 expression can be upregulated following T cell activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD148
Applications ELISA
Classification Monoclonal
Clone 10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PTPRJ
Gene Accession No. Q12913
Gene Alias AI450271; BET; Byp; CD148; CD148 antigen; colon tumor susceptibility 1; density enhanced phosphatase-1; density-enhanced phosphatase 1; DEP1; DEP-1; HPTP beta-like tyrosine phosphatase; HPTP eta; HPTPeta; human density enhanced phosphatase-1; protein tyrosine phosphatase, receptor type J; protein tyrosine phosphatase, receptor type, J; protein tyrosine phosphatase, receptor type, J polypeptide; protein-tyrosine phosphatase eta; Protein-tyrosine phosphatase receptor type J; Ptpb2; PTPbeta2; Ptprj; Receptor-type tyrosine-protein phosphatase eta; R-PTP-ETA; RPTPJ; R-PTP-J; Scc1; Scc-1; Susceptibility to colon cancer 1; susceptibility to colon cancer 1, mouse, homolog of
Gene Symbols PTPRJ
Host Species Mouse
Immunogen Recombinant Human DEP1/PTPRJ Protein (Arg997-Ala1337).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5795
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.