Learn More
Invitrogen™ CD148 Polyclonal Antibody

Description
CD148 Polyclonal Antibody for IHC (P), Western Blot
Specifications
Specifications
| Antigen | CD148 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | PTPRJ |
| Gene Accession No. | Q12913, Q64455 |
| Gene Alias | AI450271; BET; Byp; CD148; CD148 antigen; colon tumor susceptibility 1; density enhanced phosphatase-1; density-enhanced phosphatase 1; DEP1; DEP-1; HPTP beta-like tyrosine phosphatase; HPTP eta; HPTPeta; human density enhanced phosphatase-1; protein tyrosine phosphatase, receptor type J; protein tyrosine phosphatase, receptor type, J; protein tyrosine phosphatase, receptor type, J polypeptide; protein-tyrosine phosphatase eta; Protein-tyrosine phosphatase receptor type J; Ptpb2; PTPbeta2; Ptprj; Receptor-type tyrosine-protein phosphatase eta; R-PTP-ETA; RPTPJ; R-PTP-J; Scc1; Scc-1; Susceptibility to colon cancer 1; susceptibility to colon cancer 1, mouse, homolog of |
| Gene Symbols | PTPRJ |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.