Learn More
Invitrogen™ CD15 Monoclonal Antibody (HI98), PE-Cyanine7, eBioscience™

Description
The HI98 monoclonal antibody reacts with human CD15, Lewis X. This 3-fucosyl-N-acetyllactosamine carbohydrate moiety is expressed by granulocytes. Monocytes express this structure at varying degree while lymphocytes are negative. Applications Reported: This HI98 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HI98 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount ( μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Specifications
Specifications
| Antigen | CD15 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HI98 |
| Concentration | 5 μL/Test |
| Conjugate | PE-Cyanine7 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | FUT4 |
| Gene Accession No. | P22083 |
| Gene Alias | FUTIV; Lewis X; SSEA-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.