Learn More
Invitrogen™ CD150 Monoclonal Antibody (IPO-3)
Description
MA1-7626 detects SLAM from human samples. MA1-7626 has been successfully used in ELISA and flow cytometry procedures. The MA1-7626 immunogen is an activated B cell line. Store at 4°C short term. For long term storage, store at -80°C.
Specifications
Specifications
| Antigen | CD150 |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | IPO-3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | Slamf1 |
| Gene Accession No. | Q13291 |
| Gene Alias | 4933415F16; AA177906; CD150; CD150 antigen; CDw150; ESTM51; IPO-3; RGD1560634; signaling lymphocyte activation molecule; signaling lymphocytic activation molecule; signaling lymphocytic activation molecule family member 1; Slam; SLAM family member 1; Slamf1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.