Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD153 Monoclonal Antibody (RM153), eBioscience™

Catalog No. p-7168665
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-129-22 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-129-22 Supplier Invitrogen™ Supplier No. 14153185
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The RM153 monoclonal antibody reacts with mouse CD153 (CD30 ligand). This 40 kDa cell surface receptor, a type II transmembrane protein, is a member of the TNF superfamily. Interaction of CD30 with CD153 plays an important role in T cell activation/costimulation. Expression of CD153 on T cells does not appear to require costimulation through CD28 and is upregulated via TCR engagement and peaks 24-48 hours after activation. Signaling through CD153 is important in costimulation of pre-activated T cells. Applications Reported: The RM153 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining of frozen tissue sections. It has also been reported in blocking of ligand binding. (Please use Functional Grade purified RM153, Product # 16-1531, in functional assays). Applications Tested: The RM153 antibody has been tested by flow cytometric analysis of activated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

The ligand for CD30 is CD30L (CD153). The binding of CD30 to CD30L mediates pleiotropic effects including cell proliferation, activation, differentiation, and apoptotic cell death. CD30 has a critical role in the pathophysiology of Hodgkin's disease and other CD30+ lymphomas. CD30 acts as a costimulatory molecule in thymic negative selection. In addition to its expression on Hodgkin's and Reed-Sternberg cells, CD30 is also found in some non-Hodgkin's lymphomas (including Burkitt's lymphomas), virus-infected T and B cells, and on normal T and B cells after activation. In T cells, CD30 expression is present on a subset of T cells that produce Th2-type cytokines and on CD4+/CD8+ thymocytes that coexpress CD45RO and the IL4 receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD153
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Neutralization
Classification Monoclonal
Clone RM153
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Tnfsf8
Gene Accession No. P32972
Gene Alias CD153; CD153 antigen; CD30 antigen ligand; CD30 ligand; Cd30l; CD30-L; Cd30lg; CD30ligand; Tnfsf8; TNLG3A; tumor necrosis factor (ligand) superfamily member 8; tumor necrosis factor (ligand) superfamily, member 8; tumor necrosis factor ligand 3A; tumor necrosis factor ligand superfamily member 8; tumor necrosis factor superfamily member 8
Gene Symbols Tnfsf8
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21949
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.