Learn More
Invitrogen™ CD155 Monoclonal Antibody (2H7CD155), PerCP-eFluor™ 710, eBioscience™

Description
Description: The monoclonal antibody 2H7CD155 recognizes CD155 also known as the poliovirus receptor. The CD155 protein is a member of the Ig superfamily in which 4 isoforms exist; only the alpha and delta forms are expressed on the membrane of monocytes, prostate cancer cells, as well as neurectodermal and monocytic tumor cells lines. In addition, CD155 is found during mid-gastrulation during embryonic development. CD155 is thought to play a role in adhesion by interaction with the ECM component vitronectin as well as a role in NK killing of tumor cells. CD226 and CD96 are ligands for CD155. Blocking of CD155 is under therapeutic investigation for prevention of variants of poliovirus. Applications Reported: This 2H7CD155 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2H7CD155 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells or human tumor cell line U937. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrum...
Specifications
Specifications
| Antigen | CD155 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2H7CD155 |
| Concentration | 5 μL/Test |
| Conjugate | PerCP-eFluor 710 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | PVR |
| Gene Accession No. | P15151 |
| Gene Alias | 3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4 |
| Show More |
Frequently Asked Questions (FAQs)
Our options will depend on the samples you are analyzing.
If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.
For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.
Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).
As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.