Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD158a/h/g Monoclonal Antibody (HP-MA4), PerCP-Cyanine5.5, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5015824
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-158-24 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-158-24 Supplier Invitrogen™ Supplier No. 45158942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody HP-MA4 recognizes several CD158 proteins, also known as KIR2D, specifically KIR2DL1 (CD158a), KIR2DS1 (CD158h), KIR2DS3, and KIR2DS5 (CD158g). It does not recognize KIR2DL2 (CD158b1), KIR2DS2 (CD158j), KIR2DL4 (CD158d) or KIR2DS4 (CD158i) and crossreactivity to KIR2DL3 (CD158b2) and KIR2DL5 (CD158f) are unknown. CD158 proteins are members of the killer cell immunoglobulin-like receptors (KIR) family of transmembrane glycoproteins that are expressed on NK cells and a subset of T cells. Each KIR has an isoform with either a short (S) or long (L) cytoplasmic domain which transduces either an activating or inhibitory signal, respectively. The crossreactivity of the HP-MA4 antibody to KIR2DS3 and KIR2DS5 was published in 2013. Applications Reported: This HP-MA4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HP-MA4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.

Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several 'framework' genes are found in all haplotypes. The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD158a/h/g
Applications Flow Cytometry
Classification Monoclonal
Clone HP-MA4
Concentration 5 μL/Test
Conjugate PerCP-Cyanine5.5
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene KIR2DS1
Gene Accession No. P43626, P43631, P43632, Q14953, Q14954, Q99706
Gene Alias 183ActI; CD158; CD158 antigen-like family member A; CD158 antigen-like family member G; CD158 antigen-like family member H; CD158 antigen-like family member I; CD158 antigen-like family member J; CD158A; CD158b; CD158B1; CD158G; CD158H; CD158I; CD158j; cl-49; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 1; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 5; killer cell immunoglobulin-like receptor 2DL1; Killer cell immunoglobulin-like receptor 2DS1; killer cell immunoglobulin-like receptor 2DS2; Killer cell immunoglobulin-like receptor 2DS4; killer cell immunoglobulin-like receptor 2DS5; killer cell immunoglobulin-like receptor KIR2DS2; killer cell immunoglobulin-like receptor KIRDS1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; killer Ig receptor; killer inhibitory receptor 2-2-1; killer inhibitory receptor 4-1-2; killer-cell Ig-like receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor two domains short tail 2 protein; KIR antigen 2DS4; KIR1D; KIR221; KIR2DL1; KIR2DP1DL1; KIR2DS1; KIR2DS2; KIR-2DS2; KIR2DS4; KIR-2DS4; KIR2DS5; KIR412; KIR-K64; KKA3; MHC class I NK cell receptor; MHC class I NK cell receptor Eb6 ActI; natural killer associated transcript 5; natural killer cell inhibitory receptor; natural killer cell receptor; natural killer-associated transcript 1; Natural killer-associated transcript 5; Natural killer-associated transcript 8; natural killer-associated transcript 9; NK receptor 183 ActI; NKAT; NKAT1; NKAT-1; NKAT5; NKAT-5; NKAT6; NKAT8; NKAT-8; NKAT9; NKAT-9; p50 killer cell activating receptor KAR-K1d; p50.1; p50.3; p58 killer cell inhibitory receptor KIR-K64; p58 killer cell inhibitory receptor KIR-K7a; p58 natural killer cell receptor clone CL-49; P58 natural killer cell receptor clones CL-39/CL-17; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-39/CL-17; p58 NK receptor CL-42/47.11; p58 NK receptor CL-49; p58.1; p58.1 MHC class-I-specific NK receptor; XXbac-BCX195L8.1
Gene Symbols KIR2DL1, KIR2DL4, KIR2DS1, KIR2DS2, KIR2DS4, KIR2DS5
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100132285, 3802, 3805, 3806, 3809, 3810
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.