Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD158b Mouse anti-Human, PE, Clone: CH-L, BD

Catalog No. BDB561723 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561723 25 Tests
BDB559785 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561723 Supplier BD Biosciences Supplier No. 561723
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 50 - 58 kDa type I glycoprotein molecule of the killer cell inhibitory receptor (KIR) family of proteins. CD158b has also been known as GLI83, p50.2, or p58.2. CD158b is composed of two extracellular Ig-like domains. It is expressed on a subset of NK cells and it may also be present on rare T cells (either CD3/TCR αβ+, or CD3/TCR γδ+). CD158b regulates NK cell mediated cytolytic activity by interacting with HLA-C alleles (Cw 1, 3, 7 and 8).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD158b
Applications Flow Cytometry
Classification Monoclonal
Clone CH-L
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.