Learn More
Proteintech Group Inc CD16 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 165591AP150UL
Encompass_Preferred
The CD16 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD16. This antibody recognizes human antigen. The CD16 antibody has been validated for the following applications: FC, IHC, WB,ELISA analysis.
Catalog No. 50-225-6411
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.