Learn More
Invitrogen™ CD162 Polyclonal Antibody

Description
CD162 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | CD162 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SELPLG |
| Gene Accession No. | Q14242 |
| Gene Alias | CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG |
| Gene Symbols | SELPLG |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.