Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD162 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5144701
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144701 Supplier Thermo Scientific Supplier No. PA5144701

Rabbit Polyclonal Antibody

Immunogen sequence: LWDTWADEAE KALGPLLARD RRQATEYEYL DYDFLPETEP PEMLRNSTDT TPLTGPGTPE STTVEPAARR STGLDAGGAV TELTTELANM GNLSTDSAAM EIQTTQPAAT EAQTTQPVPT EAQTTPLAAT Antigen sequence identity: Mouse 34%; Rat 31%

CD162 Polyclonal antibody specifically detects CD162 in Human samples. It is validated for Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD162
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SELPLG
Gene Accession No. Q14242
Gene Alias CD162, CLA, Cutaneous lymphocyte associated antigen, cutaneous lymphocyte-associated associated antigen, leukocyte cell surface adhesion molecule, P-selectin glycoprotein ligand 1, P-selectin glycoprotein ligand 1 propeptide, P-selectin glycoprotein ligand-1, Psgl1, Psgl-1, Selectin P ligand, selectin, platelet (p-selectin) ligand, Selp1, SELPG, Selpl, SELPLG
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 20-149.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6404
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.