Learn More
Roche Diagnostics CD163 (MRQ-26) Mouse Monoclonal Antibody
Supplier:  Roche Diagnostics 5973929001
Encompass
CD163 (MRQ-26) Mouse Monoclonal Primary Antibody is intended for laboratory use in the detection of the CD163 protein in formalin-fixed, paraffin-embedded human tissue stained in qualitative immunohistochemistry (IHC) on BenchMark IHC/ISH instruments.This product should be interpreted by a qualified pathologist in conjunction with histological examination, relevant clinical information, and proper controls.This antibody is intended for in vitro diagnostic (IVD) use.
Catalog No. NC2895026
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.