Learn More
Invitrogen™ CD165 Monoclonal Antibody (SN2), PE

Description
This antibody recognizes an extracellular epitope of CD165, an approximately 37-42 kDa transmembrane glycoprotein expressed mainly on leukemic T cells, double positive and double negative thymocytes (CD4-CD8-, CD4+CD8+), and platelets. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.
Specifications
Specifications
| Antigen | CD165 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SN2 |
| Concentration | 0.08 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene Alias | gp137 |
| Host Species | Mouse |
| Immunogen | CD165, from human Molt-4 cell line. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.