Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD166 (ALCAM) Monoclonal Antibody (eBioALC48), PE, eBioscience™, Invitrogen™

Catalog No. 5010679
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-106-79 50 μg
50-106-80 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-106-79 Supplier Invitrogen™ Supplier No. 12166181
  • $168.00
    $126.65 / Each of 1

    Save $41.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: Activated leukocyte cell adhesion molecule (ALCAM; CD166) is a 105 kD cell-surface adhesion molecule belonging to the Ig superfamily of receptors. ALCAM is a type I transmembrane glycoprotein, possessing five extracellular Ig domains, a transmembrane domain, and a short cytoplasmic tail. ALCAM is involved in homotypic adhesion, as well as heterotypic adhesion with its natural ligand, CD6. A soluble isoform of ALCAM has been shown to inhibit homophilic adhesive interactions. ALCAM is expressed on a variety of cell types, including activated T and B cells, monocyte-derived dendritic cells, and thymic epithelial cells. ALCAM expression has also been used as a marker for the isolation of pluripotent stem cells. Applications Reported: This eBioALC-48 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioALC-48 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

This gene encodes activated leukocyte cell adhesion molecule (ALCAM), also known as CD166 (cluster of differentiation 166), which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. This protein binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD166 (ALCAM)
Applications Flow Cytometry
Classification Monoclonal
Clone eBioALC48
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ALCAM
Gene Accession No. Q61490
Gene Alias activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product
Gene Symbols ALCAM
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11658
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.