Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD169 (Siglec-1) Monoclonal Antibody (SER-4), eFluor™ 660, eBioscience™, Invitrogen™

Catalog No. p-7092338
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-9190 25 μg
50-112-4394 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-9190 Supplier Invitrogen™ Supplier No. 50575580
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The SER-4 monoclonal antibody reacts with mouse CD169, which is also known as Sialoadhesin and Siglec-1. CD169 is a 185-kDa member of the Siglec family of adhesion and endocytic receptors that recognize sialylated glycans. This receptor is expressed on a subset of macrophages that are found with high frequency in the spleen, liver, lymph node, and bone marrow. CD169 can also be detected on monocytes in the bone marrow and periphery. This receptor plays a role in cell-cell adhesion, as well as internalization of sialylated virus and bacteria. This endocytic pathway allows CD169+ macrophages to present lipid antigens to iNKT cells, thereby leading to their activation. Studies have also demonstrated that direct interaction of CD169 with regulatory T cells inhibits their proliferation. The SER-4 antibody has been reported to have antagonist activity. Applications Reported: This SER-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SER-4 antibody has been tested by flow cytometric anaylsis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD169 (Siglec-1)
Applications Flow Cytometry
Classification Monoclonal
Clone SER-4
Concentration 0.2 mg/mL
Conjugate eFluor 660
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene SIGLEC1
Gene Accession No. Q62230
Gene Alias Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN
Gene Symbols SIGLEC1
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20612
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
I am unable to analyze my cells stained with eFluor Organic Dyes today. What options do I have?

Our options will depend on the samples you are analyzing.

If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.

For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.

Is the eFluor 660 fluorochrome compatible with Anti-Cy5/Alexa Fluor 647 beads?

Yes, in-house studies have demonstrated that the eFluor 660 fluorochrome is recognized by Anti-Cy5/Alexa Fluor 647 beads. Side by side studies with Alexa Fluor 647 versus eFluor 660 conjugated antibodies have demonstrated comparable results.

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.