Learn More
Sino Biological Human CD16b / FCGR3B Protein (NA2 allotype, His Tag) 100ug
Supplier:  Sino Biological 11046H08C100
Encompass_Preferred
A DNA sequence encoding the human CD16b (O75015.2) (Met 1-Ser 200) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-211-6292
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.