Learn More
Invitrogen™ CD177 Monoclonal Antibody (MEM-166)

Description
This antibody reacts with CD177 (Neutrophil specific antigen 1), a 60 kDa GPI-linked cell surface glycoprotein of uPAR family, expressed on granulocytes and in bone marrow early erythroblasts, megakaryocytes, promyelocytes and myelocytes. Western blot should be performed under non-reducing conditions.
Specifications
Specifications
| Antigen | CD177 |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | MEM-166 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | CD177 |
| Gene Accession No. | Q8N6Q3 |
| Gene Alias | 1190003K14Rik; CD177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2a; Human neutrophil alloantigen 2a; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.