Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD181 (CXCR1) Rat anti-Mouse, PE, Clone: U45-632, BD

Catalog No. BDB566383 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566383 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566383 Supplier BD Biosciences Supplier No. 566383
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The U45-632 monoclonal antibody specifically binds to mouse CD181, also known as C-X-C chemokine receptor type 1 (CXCR1) or IL-8 receptor . CD181 is a chemokine receptor of the G-protein coupled receptor 1 family, also known as seven-transmembrane domain receptors. In the mouse, which is not known to express IL-8 (CXCL8), CD181 binds to mouse GCP-2 (CXCL6). Mouse CD181 can also bind to human IL-8. CD181 is expressed by neutrophils, which are activated upon ligand binding. The chemokine receptors CXCR1 and CXCR2 play an important role in mediating neutrophil recruitment and neutrophil-dependent injury in several models of inflammation.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD181 (CXCR1)
Classification Monoclonal
Clone U45-632
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Il8ra,Cxcr1,C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, IL-8R A
Host Species Rat
Immunogen Mouse CD181 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.