Learn More
Sino Biological Recombinant Human CD19 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 11880H02H50
Encompass_Preferred
A DNA sequence encoding the human CD19 (NP_001171569.1) (Met1-Lys291) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. NC1710120
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.