Learn More
Sino Biological Anti-CD19 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90051T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus / Rhesus CD19 ( Catalog#90051-C08H; XP_005591597.1; Met1-Lys292). CD19 specific IgG was purified by Cynomolgus / Rhesus CD19 affinity chromatography.
Catalog No. 50-228-3297
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.