Learn More
Invitrogen™ CD19 Monoclonal Antibody (CB19)
Description
MA1-7638 detects CD19 from human samples. MA1-7638 has been successfully used in flow cytometry, fluorescent microscopy, and Immunohistology (Frozen) procedures. The MA1-7638 immunogen is a CD19 protein.
Specifications
Specifications
| Antigen | CD19 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | CB19 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.01% sodium azide; pH 7.2 |
| Gene | Cd19 |
| Gene Accession No. | P15391 |
| Gene Alias | AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.