Learn More
Beckman Coulter CD19, Single Color Antibody, IM2643U
Supplier:  Beckman Coulter A66328
Encompass
The Beckman Coulter CD19 antibody IM2643U is a purified mouse monoclonal antibody conjugated to R-phycoerythrin (RPE). It is designed for flow cytometry analysis of human CD19, a 95 kDa transmembrane glycoprotein expressed on B lymphocytes and follicular dendritic cells. This antibody can be used for immunophenotyping of leukemias and lymphomas and monitoring B cell levels. It is supplied as a liquid in sodium azide at 0.1% concentration. The recommended usage is 5-10 μL per test, with 106 cells per test. It has a 12 month shelf life when stored at 2-8°C protected from prolonged exposure to light.
Catalog No. NC1909489
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.