Learn More
Beckman Coulter CD19-PC7, J3-119, 1 mL, ASR
Supplier:  Beckman Coulter IM3628U
Encompass
The CD19 antigen (also called B4) is a type I membrane glycoprotein with a molecular weight of 95 kDa. CD19 is involved in the regulation of B lymphocyte development, activation and differentiation.
Catalog No. NC9842644
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.