Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD196 (CCR6) Monoclonal Antibody (R6H1), FITC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 509781
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-978-1 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-978-1 Supplier Invitrogen™ Supplier No. 11196942
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This R6H1 monoclonal antibody reacts with CD196 (also known as CCR6), a seven transmembrane G protein-coupled receptor expressed on T, B, dendritic, natural killer, and Langerhans cells. This CC chemokine receptor uniquely binds MIP-3a/CCL20, a chemoattractant for dendritic cells, effector/memory T cells, and B cells. CD196 is also involved in host defense and inflammation at epithelial sites. Furthermore, this receptor has been implicated in Th17 differentiation and CD4+FoxP3+ regulatory T cell development. Applications Reported: This R6H1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This R6H1 antibody has been pre-titrated and tested by flow cytometric analysis on normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD196, also known as CCR6, is a member of the beta chemokine receptor family and is characterized by a seven transmembrane structure similar to G protein-coupled receptors. This receptor uniquely binds to macrophage inflammatory protein 3 alpha (MIP-3alpha/CCL20), which acts as a chemoattractant for dendritic cells, effector/memory T cells, and B cells. CCR6 is preferentially expressed by immature dendritic cells and memory T cells, playing a crucial role in host defense and inflammation at epithelial sites. It is important for B-lineage maturation and antigen-driven B-cell differentiation, and it regulates the migration and recruitment of dendritic and T cells during inflammatory and immunological responses. Additionally, CCR6 has been implicated in the differentiation of Th17 cells and the development of CD4+FoxP3+ regulatory T cells, highlighting its role in immune regulation. Alternatively spliced transcript variants encoding the same protein have been described for this gene, further emphasizing its functional diversity in immune processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD196 (CCR6)
Applications Flow Cytometry
Classification Monoclonal
Clone R6H1
Concentration 5 μL/Test
Conjugate FITC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CCR6
Gene Accession No. P51684
Gene Alias BN-1; CC chemokine receptor 6; C-C chemokine receptor type 6; C-C CKR-6; C-C motif chemokine receptor 6; CC-CKR-6; CCR6; CCR-6; CD196; chemokine (C-C motif) receptor 6; chemokine (C-C) receptor 6; chemokine receptor-like 3; CKR6; CKRL3; CKR-L3; Cmkbr6; DCR2; DRY6; DRY-6; G protein-coupled receptor 29; GPR29; GPRCY4; GPR-CY4; G-protein coupled receptor 29; KY411; LARC receptor; seven-transmembrane receptor, lymphocyte, 22; STRL22
Gene Symbols CCR6
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1235
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.