Learn More
Invitrogen™ CD197 (CCR7) Monoclonal Antibody (3D12), Brilliant Ultra Violet™ 563, eBioscience™, Invitrogen™

Description
Specifications
Specifications
| Antigen | CD197 (CCR7) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 3D12 |
| Concentration | 5 μL/Test |
| Conjugate | Brilliant Ultraviolet 563 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Ccr7 |
| Gene Accession No. | P32248 |
| Gene Alias | BLR2; Bukitt's lymphoma receptor 2; CC chemokine receptor 7; C-C chemokine receptor type 7; C-C CKR-7; C-C motif chemokine receptor 7; CC-CKR-7; CCR7; CCR-7; CD197; CDw197; chemokine (C-C motif) receptor 7; chemokine (C-C) receptor 7; Cmkbr7; EBI1; Ebi1h; EBV-induced G protein-coupled receptor 1; EBV-induced G-protein coupled receptor 1; Epstein-Barr virus induced gene 1; Epstein-Barr virus-induced G-protein coupled receptor 1; EVI1; lymphocyte-specific G protein-coupled peptide receptor; MIP-3 beta receptor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.