Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD1w2 Monoclonal Antibody (CC20), PE

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528306
Description
Reconstitute with 1 mL distilled water.
CD1w2 gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to the plasma membrane and to recycling vesicles of the early endocytic system. Alternatively spliced transcript variants have been observed, but their biological validity has not been determined.
Specifications
| CD1w2 | |
| Monoclonal | |
| Conc. Not Determined | |
| PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
| 0 | |
| CD1A | |
| Protein A | |
| RUO | |
| 781011 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
| Flow Cytometry | |
| CC20 | |
| PE | |
| CD1A | |
| CD1A; CD1a molecule; T-cell surface glycoprotein CD1a | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction