Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD2/CD20 Antibody Cocktail, FITC, PE
GREENER_CHOICE

Catalog No. 01670781 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-781 100 Tests
01-670-782 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-670-781 Supplier Invitrogen™ Supplier No. MA112058
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody Cocktail

This antibody is a monoclonal mixture of FITC-conjugated CD2 (clone T6.3, isotype IgG2a) and PE-conjugated CD20 (clone DFA-7, isotype IgG1).

CD2 is found on the majority of human T and NK cells. CD2 presents on 78+8% normal peripheral lymphocytes and 100% of thymocytes. This antibody detects CD2 antigen CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lamda light chains. It appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and malignant B cell proliferation. It is expressed in all B cell precursor leukemias.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2/CD20
Applications Flow Cytometry
Classification Cocktail
Clone T6.3, DFA-7, DFA-7
Conjugate FITC/PE
Formulation PBS with 0.2% carrier protein and 0.08% sodium azide
Gene MS4A1
Gene Accession No. P06729, P11836
Gene Alias 2CD2 antigen; B1; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; CD2; CD2 molecule; CD20; CD20 antigen; CD20 receptor; CVID5; Erythrocyte receptor; LEU-16; leukocyte surface antigen Leu-16; LFA-2; LFA-3 receptor; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; MGC3969; MS4A1; MS4A2; Rosette receptor; S7; SRBC; T11; T-cell surface antigen CD2; T-cell surface antigen T11/Leu-5
Gene Symbols CD2, MS4A1
Host Species Mouse
Immunogen T lymphocytes activated by mixed lymphocyte culture and the LB lymphoblastoid cell line.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 914, 931
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1, IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.