Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD2/CD7 Antibody Cocktail, FITC, PE
GREENER_CHOICE

Catalog No. 01670727 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-727 100 Tests
01-670-728 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-670-727 Supplier Invitrogen™ Supplier No. MA112004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody Cocktail

This antibody is a monoclonal mixture of FITC-conjugated CD2 (clone T6.3, isotype IgG2a) and PE-conjugated CD7 (clone MG34, isotype IgG2a).

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2/CD7
Applications Flow Cytometry
Classification Cocktail
Clone T6.3, MG34, MG34
Conjugate FITC/PE
Formulation PBS with 0.2% carrier protein and 0.08% sodium azide
Gene CD7
Gene Accession No. P06729, P09564
Gene Alias 2CD2 antigen; CD2; CD2 molecule; CD7; CD7 antigen (p41); CD7 molecule; Erythrocyte receptor; GP40; LEU-9; LFA-2; LFA-3 receptor; p41 protein; Rosette receptor; SRBC; T11; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen CD2; T-cell surface antigen Leu-9; T-cell surface antigen T11/Leu-5; Tp40; TP41
Gene Symbols CD2, CD7
Host Species Mouse
Immunogen T lymphocytes activated by mixed lymphocyte culture and T-acute lymphoblastic leukemia (T-ALL) cells.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 914, 924
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.