Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD200R3 Monoclonal Antibody (Ba13), PE-Cyanine7, eBioscience™, Invitrogen™

Catalog No. p-7192863
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-200-180 25 μg
25-200-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-200-180 Supplier Invitrogen™ Supplier No. 25200180
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: This Ba13 monoclonal antibody recognizes mouse CD200R3, a surface receptor expressed by basophils and mast cells. Applications Reported: This Ba13 antibody has been reported for use in flow cytometric analysis. Applications Tested: This Ba13 antibody has been tested by flow cytometric analysis of the MC/9 cell line. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

CD200R3 is a dimeric cell surface receptor also known as CD200RLb. CD200R3 contains two Ig-like extracellular domains and as such is a member of the immunoglobulin superfamily of receptors. The intracellular region of CD200R3 associates with the adaptor molecule DAP12, and possibly also DAP10, CD3 zeta and FcR gamma-chain, indicating that it is capable of transducing stimulatory signals. In this respect CD200R3 bears similarity to activatory receptors CD200R2 and CD200R4. In contrast, an inhibitory receptor CD200R (also known as CD200R1) relies on other adaptors. CD200R3 is primarily expressed by basophils and mast cells, cells known to be involved in anti-parasitic immune responses and frequently also allergic reactions. The surface expression of CD200R3 decreases when basophils are activated by ligating their CD16 and CD32. Remarkably, most of this downregulation is observed within the first 30 minutes after activation. Unlike CD200R, CD200R3 does not bind CD200.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200R3
Applications Flow Cytometry
Classification Monoclonal
Clone Ba13
Concentration 0.2 mg/mL
Conjugate PE-Cyanine7
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CD200R3
Gene Accession No. Q5UKY4
Gene Alias 4733401I18Rik; 4833409J19Rik; AI505817; BB106877; CD200 cell surface glycoprotein receptor 3; CD200 cell surface glycoprotein receptor-like 3; CD200 cell surface glycoprotein receptor-like b; CD200 receptor 3; CD200 receptor-like 3; Cd200r3; CD200RLb; cell surface glycoprotein CD200 receptor 3; Cell surface glycoprotein OX2 receptor 3; mCD200RLb
Gene Symbols CD200R3
Host Species Rat
Immunogen Mouse primary basophils.
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74603
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Product Line eBioscience
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.