Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD201 (EPCR) Monoclonal Antibody (RCR-227), APC, eBioscience™
GREENER_CHOICE

Rat Monoclonal Antibody

Supplier:  Invitrogen™ 17201842

Encompass_Preferred

 View more versions of this product

Catalog No. 50-112-3100


Only null left
Edge
Add to Cart

Description

Description

Description: The monoclonal antibody RCR-227 reacts with human CD201, which is also known as the endothelial protein C receptor (EPCR), a 46-kDa transmembrane protein related to class I MHC. CD201 was originally identified on vascular endothelium cells, predominantly larger blood vessels. It is also expressed on monocytes, lymphocytes, hematopoietic stem cells, neutrophils, and eosinophils. CD201 serves as a binding site for protein C, activated protein C (APC), and factor VII/VIIa. A soluble form has also been identified in the plasma. CD201 plays a role in controlling coagulation as disruption of its gene in mice caused placental thrombosis and early embryonic death. This cell surface molecule is also involved in the anti-inflammatory response. APC binding to CD201 enables PAR-1 activation that confers cytoprotective effects. In monocytes, APC binding decreases secretion of proinflammatory cytokines, chemokines, and growth factors. Finally, CD201 has been shown to inhibit lymphocyte migration. Applications Reported: This RCR-227 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RCR-227 antibody has been pre-titrated and tested by flow cytometric analysis of HUVECs. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-...

The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD201 (EPCR)
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Q9UNN8
Procr
Affinity chromatography
RUO
10544
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
RCR-227
APC
Procr
Activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial
Rat
100 Tests
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.