Learn More
Invitrogen™ CD203a Monoclonal Antibody (PM18-7), FITC

Description
This antibody recognizes porcine CD203a, originally clustered as SWC9 at the Second International Swine CD Workshop and later identified as the porcine homologue of human ecto-nucleotidepyrophosphatase/phosphodiesterase 1 or ENPP1. SWC1a, expressed at very much higher levels on monocytes than mature macrophages and CD203a (SWC9), expressed exclusively on mature tissue macrophages have been used as markers of monocyte-macrophage differentiation.
Specifications
Specifications
| Antigen | CD203a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | PM18-7 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 1% BSA and 0.09% sodium azide |
| Gene Alias | CD203; SWC9 |
| Host Species | Mouse |
| Immunogen | Porcine alveolar macrophages. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.